The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008221.4 | NP_032247.1 | 1 - 2 | 148 | 95 |
GenBank mRNA | AK011619.1 | - | 1 - 2 | 149 | 95 |
AK012501.1 | - | 1 - 2 | 150 | 95 | |
AK012897.1 | - | 1 - 2 | 149 | 95 | |
BC057014.1 | - | 1 - 2 | 125 | 95 | |
M26897.1 | - | 1 - 2 | 110 | 95 |