The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008308.4 | NP_032334.2 | 1 - 1 | 2540 | 99 |
GenBank mRNA | AK034017.1 | - | 1 - 1 | 1988 | 99 |
AK043668.1 | - | 1 - 1 | 1999 | 99 | |
AK049814.1 | - | 1 - 1 | 2380 | 99 | |
AK049884.1 | - | 1 - 1 | 2471 | 99 | |
AK140194.1 | - | 1 - 1 | 2594 | 99 | |
BC138669.1 | - | 1 - 1 | 2247 | 99 | |
BC138681.1 | - | 1 - 1 | 2247 | 99 | |
U39391.1 | - | 1 - 1 | 2049 | 99 |