Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Il10ra
Entrez Gene ID
16154
Gene Name
interleukin 10 receptor, alpha
Gene Aliases
AW553859, CDw210, CDw210a, Il10r, mIL-10R
Chromosome Location
Chr.9: 45253837 - 45269251 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008348.2 NP_032374.1 2 - 3 267 87
GenBank mRNA AK137663.1 - 2 - 3 268 87
AK149680.1 - 2 - 3 262 87
AK150022.1 - 2 - 3 264 87
AK150524.1 - 2 - 3 271 87
AK153120.1 - 2 - 3 271 87
AK159566.1 - 2 - 3 264 87
L12120.1 - 2 - 3 272 87

View Full Product Details