The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008349.5 | NP_032375.2 | 1 - 2 | 137 | 102 |
GenBank mRNA | AF440787.1 | - | 1 - 2 | 64 | 102 |
AK040938.1 | - | 1 - 2 | 53 | 102 | |
AK134042.1 | - | 1 - 2 | 130 | 102 | |
BC145791.1 | - | 1 - 2 | 137 | 102 | |
U53696.1 | - | 1 - 2 | 50 | 102 |