Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ltf
Entrez Gene ID
17002
Gene Name
lactotransferrin
Gene Aliases
Csp82, Lf, MMS10R, Ms10r
Chromosome Location
Chr.9: 111019292 - 111042766 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008522.3 NP_032548.2 2 - 3 235 68
GenBank mRNA AK036491.1 - 2 - 3 236 68
AK144556.1 - 2 - 3 236 68
AK151822.1 - 2 - 3 236 68
AK164781.1 - 2 - 3 237 68
AY792500.1 - 2 - 3 201 68
BC006904.1 - 2 - 3 243 68
BC008530.1 - 2 - 3 216 68
CT010339.1 - 2 - 3 201 68
D88510.1 - 2 - 3 235 68
FJ538998.1 - 2 - 3 201 68
J03298.1 - 2 - 3 204 68

View Full Product Details