The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111073.2 | NP_001104543.1 | 4 - 5 | 681 | 73 |
NM_001287213.1 | NP_001274142.1 | 5 - 6 | 553 | 73 | |
NM_001287217.1 | NP_001274146.1 | 5 - 6 | 544 | 73 | |
NM_008761.4 | NP_032787.1 | 5 - 6 | 541 | 73 | |
GenBank mRNA | AI121044.1 | - | 5 - 6 | 541 | 73 |
AK003068.1 | - | 5 - 6 | 324 | 73 | |
AK155763.1 | - | 4 - 5 | 661 | 73 | |
AK169966.1 | - | 5 - 6 | 324 | 73 | |
AK170810.1 | - | 5 - 6 | 329 | 73 | |
BC031112.1 | - | 5 - 6 | 313 | 73 | |
U72680.1 | - | 5 - 6 | 347 | 73 |