The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146268.1 | NP_001139740.1 | 22 - 23 | 3560 | 87 |
NM_008809.2 | NP_032835.2 | 22 - 23 | 3557 | 87 | |
GenBank mRNA | AK134785.1 | - | 23 - 24 | 3582 | 87 |
AK143247.1 | - | 22 - 23 | 3539 | 87 | |
BC055311.1 | - | 22 - 23 | 3572 | 87 | |
DQ979390.1 | - | 16 - 17 | 2213 | 87 | |
X04367.1 | - | 22 - 23 | 3266 | 87 |