The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008887.2 | NP_032913.1 | 1 - 2 | 401 | 89 |
GenBank mRNA | CJ058587.1 | - | 1 - 2 | 402 | 89 |
CJ133172.1 | - | 1 - 2 | 151 | 89 | |
X75014.1 | - | 1 - 2 | 243 | 89 |