The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253890.1 | NP_001240819.1 | 7 - 8 | 838 | 71 |
NM_008923.3 | NP_032949.3 | 7 - 8 | 840 | 71 | |
GenBank mRNA | AK161621.1 | - | 7 - 8 | 771 | 71 |
CO425808.1 | - | 3 - 4 | 253 | 71 | |
M20473.1 | - | 7 - 8 | 786 | 71 |