The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008964.4 | NP_032990.1 | 1 - 2 | 1697 | 73 |
GenBank mRNA | AK036531.1 | - | 1 - 2 | 1615 | 73 |
AK054132.1 | - | 1 - 2 | 1218 | 73 | |
AK086904.1 | - | 1 - 2 | 1612 | 73 | |
AK136577.1 | - | 1 - 2 | 710 | 73 | |
AK137515.1 | - | 1 - 2 | 907 | 73 | |
BC005440.1 | - | 1 - 2 | 1205 | 73 | |
D50589.1 | - | 1 - 2 | 988 | 73 |