The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009059.2 | NP_033085.2 | 11 - 12 | 1541 | 73 |
GenBank mRNA | AK144440.1 | - | 11 - 12 | 1543 | 73 |
BC068121.1 | - | 11 - 12 | 1541 | 73 | |
KJ650210.1 | - | 10 - 11 | 1349 | 73 | |
U54639.1 | - | 11 - 12 | 1470 | 73 |