The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009288.2 | NP_033314.2 | 18 - 19 | 2902 | 71 |
GenBank mRNA | AK148118.1 | - | 18 - 19 | 2842 | 71 |
AK220448.1 | - | 18 - 19 | 2824 | 71 | |
BC069840.1 | - | 18 - 19 | 2975 | 71 | |
BC096624.1 | - | 18 - 19 | 2987 | 71 | |
BC128363.1 | - | 18 - 19 | 2788 | 71 | |
D89728.1 | - | 18 - 19 | 2863 | 71 |