The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001173505.1 | NP_001166976.1 | 3 - 4 | 372 | 92 |
NM_001289894.1 | NP_001276823.1 | 3 - 4 | 372 | 92 | |
NM_001289896.1 | NP_001276825.1 | 3 - 4 | 372 | 92 | |
NM_009379.3 | NP_033405.1 | 3 - 4 | 526 | 92 | |
GenBank mRNA | AI314551.1 | - | 3 - 4 | 372 | 92 |
AK047512.1 | - | 3 - 4 | 527 | 92 | |
BC003803.1 | - | 3 - 4 | 638 | 92 | |
L34169.1 | - | 3 - 4 | 308 | 92 |