Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnnc1
Entrez Gene ID
21924
Gene Name
troponin C, cardiac/slow skeletal
Gene Aliases
AI874626, TnC, cTnC, cTnI, tncc
Chromosome Location
Chr.14: 31208312 - 31211711 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009393.2 NP_033419.1 4 - 5 364 83
GenBank mRNA BC061172.1 - 4 - 5 358 83
M29793.1 - 4 - 5 364 83

View Full Product Details