Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Trpc5
Entrez Gene ID
22067
Gene Name
transient receptor potential cation channel, subfamily C, member 5
Gene Aliases
CCE2, TRP-5, TRP5, Trrp5
Chromosome Location
Chr.X: 144377614 - 144688180 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009428.2 NP_033454.1 4 - 5 1633 80
GenBank mRNA AF029983.1 - 4 - 5 1288 80
AF060107.1 - 4 - 5 1632 80
AJ006204.1 - 3 - 4 1230 80
AK038531.1 - 4 - 5 1457 80
AK047867.1 - 2 - 3 654 80
BC112972.1 - 4 - 5 1451 80

View Full Product Details