The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009701.4 | NP_033831.1 | 3 - 4 | 856 | 106 |
GenBank mRNA | AF087654.1 | - | 3 - 4 | 1105 | 106 |
AK009302.1 | - | 3 - 4 | 741 | 106 | |
BC026379.1 | - | 2 - 3 | 1050 | 106 | |
BC150769.1 | - | 4 - 5 | 902 | 106 |