The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010051.3 | NP_034181.2 | 2 - 3 | 534 | 62 |
GenBank mRNA | AF030433.1 | - | 2 - 3 | 453 | 62 |
AK141693.1 | - | 2 - 3 | 558 | 62 | |
AK144119.1 | - | 2 - 3 | 560 | 62 | |
BC050189.1 | - | 2 - 3 | 534 | 62 | |
JN966751.1 | - | 2 - 3 | 587 | 62 |