Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Jak3
Entrez Gene ID
16453
Gene Name
Janus kinase 3
Gene Aliases
fae, wil
Chromosome Location
Chr.8: 71676383 - 71690577 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190830.1 NP_001177759.1 4 - 5 631 78
NM_010589.6 NP_034719.2 4 - 5 631 78
GenBank mRNA AK038268.1 - 3 - 4 442 78
AK088365.1 - 4 - 5 483 78
AK142178.1 - 4 - 5 469 78
AK156646.1 - 4 - 5 453 78
AK164520.1 - 4 - 5 471 78
BC027234.1 - - 78
BC105577.1 - 3 - 4 434 78
BC131646.1 - 3 - 4 442 78
BC131647.1 - 3 - 4 442 78
L32955.1 - 3 - 4 433 78
L33768.1 - 3 - 4 427 78
L40172.1 - 4 - 5 492 78

View Full Product Details