The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010694.1 | NP_034824.1 | 4 - 5 | 419 | 98 |
GenBank mRNA | BC116816.1 | - | 4 - 5 | 440 | 98 |
BC116818.1 | - | 4 - 5 | 440 | 98 | |
D38580.1 | - | 4 - 5 | 419 | 98 |