Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lmx1b
Entrez Gene ID
16917
Gene Name
LIM homeobox transcription factor 1 beta
Gene Aliases
LMX1.1, LMX1.2
Chromosome Location
Chr.2: 33560965 - 33640992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010725.2 NP_034855.2 1 - 2 70 133
GenBank mRNA AB221651.1 - 1 - 2 70 133
AF078166.1 - 1 - 2 70 133
BC119169.1 - 1 - 2 149 133
BC125469.1 - 1 - 2 149 133

View Full Product Details