The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011057.3 | NP_035187.2 | 4 - 5 | 1268 | 62 |
GenBank mRNA | AF162784.1 | - | 4 - 5 | 458 | 62 |
BC023427.1 | - | 3 - 4 | 392 | 62 | |
BC053430.1 | - | 4 - 5 | 1268 | 62 | |
BC064056.1 | - | 4 - 5 | 935 | 62 |