The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205214.1 | NP_001192143.1 | 4 - 5 | 972 | 72 |
NM_001205215.1 | NP_001192144.1 | 4 - 5 | 695 | 72 | |
NM_001205216.1 | NP_001192145.1 | 4 - 5 | 695 | 72 | |
NM_011306.4 | NP_035436.1 | 4 - 5 | 972 | 72 | |
GenBank mRNA | AK140371.1 | - | 4 - 5 | 711 | 72 |
AK159671.1 | - | 4 - 5 | 973 | 72 | |
BC019432.1 | - | 4 - 5 | 627 | 72 | |
BC049773.1 | - | 4 - 5 | 932 | 72 | |
KJ650219.1 | - | 4 - 5 | 786 | 72 | |
KJ663717.1 | - | 4 - 5 | 786 | 72 | |
KJ663718.1 | - | 5 - 6 | 672 | 72 | |
KJ663719.1 | - | 5 - 6 | 672 | 72 | |
KJ663720.1 | - | 4 - 5 | 603 | 72 | |
KJ663721.1 | - | 5 - 6 | 657 | 72 | |
M26804.1 | - | 3 - 4 | 564 | 72 | |
M84818.1 | - | 4 - 5 | 645 | 72 | |
X66224.1 | - | 3 - 4 | 564 | 72 |