The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272024.1 | NP_001258953.1 | 4 - 5 | 607 | 57 |
NM_011351.2 | NP_035481.1 | 1 - 2 | 175 | 57 | |
NR_073549.1 | 2 - 3 | 454 | 57 | ||
NR_073550.1 | 2 - 3 | 468 | 57 | ||
NR_073551.1 | 3 - 4 | 517 | 57 | ||
GenBank mRNA | AB013729.1 | - | 1 - 2 | 140 | 57 |
AF363972.1 | - | 1 - 2 | 208 | 57 | |
AF363973.1 | - | 1 - 2 | 208 | 57 | |
AK031131.1 | - | 2 - 3 | 366 | 57 | |
AK165085.1 | - | 2 - 3 | 381 | 57 | |
BC051979.1 | - | 3 - 4 | 423 | 57 | |
BC059007.1 | - | 4 - 5 | 522 | 57 | |
BC083186.1 | - | 2 - 3 | 386 | 57 |