Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc30a4
Entrez Gene ID
22785
Gene Name
solute carrier family 30 (zinc transporter), member 4
Gene Aliases
Znt4, lm, znT-4
Chromosome Location
Chr.2: 122681233 - 122702663 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011774.3 NP_035904.2 2 - 3 784 118
GenBank mRNA AF003747.1 - 2 - 3 585 118
AK134245.1 - 2 - 3 762 118
BC117997.1 - 2 - 3 642 118

View Full Product Details