The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011881.3 | NP_036011.3 | 1 - 2 | 736 | 86 |
GenBank mRNA | BC075722.1 | - | 1 - 2 | 711 | 86 |
BC096611.1 | - | 1 - 2 | 736 | 86 |