Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Hsph1
Entrez Gene ID
15505
Gene Name
heat shock 105kDa/110kDa protein 1
Gene Aliases
105kDa, AI790491, Hsp105, Hsp110, hsp-E7I, hsp110/105
Chromosome Location
Chr.5: 149616843 - 149637155 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013559.2 NP_038587.2 15 - 16 2330 87
GenBank mRNA AK075731.1 - 5 - 6 553 87
AK083179.1 - - 2191 87
AK146697.1 - 15 - 16 2249 87
AK165046.1 - 15 - 16 2298 87
AK172913.1 - 9 - 10 1227 87
BC018378.1 - 15 - 16 2205 87
D67016.1 - 15 - 16 2145 87
D67017.1 - 14 - 15 2100 87
L40406.1 - 15 - 16 2233 87

View Full Product Details