The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139509.1 | NP_001132981.1 | 5 - 6 | 1455 | 85 |
NM_013613.2 | NP_038641.1 | 5 - 6 | 1601 | 85 | |
GenBank mRNA | AB014889.1 | - | 5 - 6 | 1194 | 85 |
AK087495.1 | - | 6 - 7 | 1479 | 85 | |
AK158607.1 | - | 5 - 6 | 1602 | 85 | |
BC137715.1 | - | 5 - 6 | 1913 | 85 | |
BC144854.1 | - | 5 - 6 | 2129 | 85 | |
S53744.1 | - | 5 - 6 | 1492 | 85 |