Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tec
Entrez Gene ID
21682
Gene Name
tec protein tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.5: 72755718 - 72868740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113460.2 NP_001106931.1 10 - 11 977 70
NM_001113461.2 NP_001106932.1 9 - 10 911 70
NM_001113464.2 NP_001106935.1 9 - 10 890 70
NM_013689.5 NP_038717.2 9 - 10 870 70
GenBank mRNA AK139329.1 - 9 - 10 886 70
AK154457.1 - 10 - 11 973 70
AK168452.1 - 9 - 10 908 70
AK168460.1 - 9 - 10 907 70
AK170588.1 - 9 - 10 909 70
BC018230.1 - 8 - 9 875 70
BC037071.1 - 9 - 10 905 70
BC062884.1 - 5 - 6 453 70
S53716.1 - 9 - 10 901 70
X55663.1 - 9 - 10 905 70

View Full Product Details