Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tec
Entrez Gene ID
21682
Gene Name
tec protein tyrosine kinase
Gene Aliases
-
Chromosome Location
Chr.5: 72755718 - 72868740 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113460.2 NP_001106931.1 12 - 13 1191 95
NM_001113461.2 NP_001106932.1 11 - 12 1125 95
NM_001113464.2 NP_001106935.1 11 - 12 1104 95
NM_013689.5 NP_038717.2 11 - 12 1084 95
GenBank mRNA AK139329.1 - 11 - 12 1100 95
AK168452.1 - 11 - 12 1122 95
AK168460.1 - 11 - 12 1121 95
AK170588.1 - 11 - 12 1123 95
BC018230.1 - 10 - 11 1089 95
BC037071.1 - 11 - 12 1119 95
BC062884.1 - 7 - 8 667 95
S53716.1 - 11 - 12 1115 95
X55663.1 - 11 - 12 1119 95

View Full Product Details