Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gosr1
Entrez Gene ID
53334
Gene Name
golgi SNAP receptor complex member 1
Gene Aliases
AI414660, AI426320, BB145494, GOS-28, GOSRI, GS28
Chromosome Location
Chr.11: 76726602 - 76763622 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016810.3 NP_058090.2 7 - 8 552 119
GenBank mRNA AF079901.1 - 7 - 8 575 119
AK133836.1 - 7 - 8 562 119
AK134515.1 - 7 - 8 555 119
AK147799.1 - 7 - 8 576 119
AK154972.1 - 7 - 8 578 119
AK161386.1 - 7 - 8 566 119
BC008542.1 - 7 - 8 552 119
BC051661.1 - 7 - 8 571 119

View Full Product Details