Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syt7
Entrez Gene ID
54525
Gene Name
synaptotagmin VII
Gene Aliases
AI851541, B230112P13Rik
Chromosome Location
Chr.19: 10388902 - 10453181 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018801.3 NP_061271.1 2 - 3 533 62
NM_173067.3 NP_775090.1 2 - 3 533 62
NM_173068.2 NP_775091.2 2 - 3 533 62
GenBank mRNA AB026804.1 - 2 - 3 140 62
AB075900.1 - 2 - 3 140 62
AB075901.1 - 2 - 3 140 62
BC105660.1 - 2 - 3 139 62
BC139806.1 - 2 - 3 140 62

View Full Product Details