The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019752.3 | NP_062726.3 | 6 - 7 | 1258 | 100 |
GenBank mRNA | AF164513.1 | - | 6 - 7 | 1137 | 100 |
AF175324.1 | - | 6 - 7 | 1154 | 100 | |
AK011536.1 | - | - | 2064 | 100 | |
AK146338.1 | - | 6 - 7 | 1149 | 100 | |
AK159188.1 | - | 6 - 7 | 1078 | 100 | |
BC033276.1 | - | 6 - 7 | 1118 | 100 | |
BC049880.1 | - | 6 - 7 | 1258 | 100 | |
BM899329.1 | - | 2 - 3 | 100 |