The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019794.4 | NP_062768.1 | 8 - 9 | 1066 | 129 |
GenBank mRNA | AB028853.1 | - | 8 - 9 | 1067 | 129 |
AK031858.1 | - | 8 - 9 | 1133 | 129 | |
AK077672.1 | - | 8 - 9 | 1137 | 129 | |
AK083208.1 | - | 8 - 9 | 1134 | 129 | |
AK163255.1 | - | 8 - 9 | 1065 | 129 | |
AK169169.1 | - | 8 - 9 | 1126 | 129 | |
BC003420.1 | - | 8 - 9 | 1139 | 129 |