The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164352.1 | NP_001157824.1 | 6 - 7 | 1059 | 94 |
NM_021474.3 | NP_067449.3 | 7 - 8 | 818 | 94 | |
GenBank mRNA | AF104223.1 | - | 6 - 7 | 773 | 94 |
AF109122.1 | - | 6 - 7 | 1032 | 94 | |
AK075585.1 | - | 7 - 8 | 807 | 94 | |
BC012269.1 | - | 6 - 7 | 956 | 94 |