The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163487.1 | NP_001156959.1 | 17 - 18 | 1801 | 90 |
NM_001163488.1 | NP_001156960.1 | 17 - 18 | 1737 | 90 | |
NM_021514.4 | NP_067489.3 | 16 - 17 | 1754 | 90 | |
GenBank mRNA | AF249894.1 | - | 16 - 17 | 1694 | 90 |
AK002711.2 | - | 17 - 18 | 1704 | 90 | |
AK049773.1 | - | 17 - 18 | 1709 | 90 | |
BC005526.1 | - | 16 - 17 | 1718 | 90 |