The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021538.1 | NP_067513.1 | 3 - 4 | 311 | 67 |
GenBank mRNA | AB039837.1 | - | 3 - 4 | 311 | 67 |
AK003458.1 | - | 3 - 4 | 328 | 67 | |
BC009170.1 | - | 3 - 4 | 319 | 67 | |
BC083336.1 | - | 3 - 4 | 293 | 67 |