Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddr2
Entrez Gene ID
18214
Gene Name
discoidin domain receptor family, member 2
Gene Aliases
AW495251, Ntrkr3, tyro10
Chromosome Location
Chr.1: 169972307 - 170110836 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022563.2 NP_072075.2 9 - 10 1339 75
GenBank mRNA BC138826.1 - 10 - 11 1585 75
BC138827.1 - 10 - 11 1480 75
G49238.1 - 10 - 11 1646 75
X76505.1 - 10 - 11 1646 75

View Full Product Details