Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ddr2
Entrez Gene ID
18214
Gene Name
discoidin domain receptor family, member 2
Gene Aliases
AW495251, Ntrkr3, tyro10
Chromosome Location
Chr.1: 169972307 - 170110836 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022563.2 NP_072075.2 12 - 13 1901 77
GenBank mRNA AK132504.1 - 9 - 10 1400 77
BC138826.1 - 13 - 14 2147 77
BC138827.1 - 13 - 14 2042 77
G49238.1 - 13 - 14 2208 77
X76505.1 - 13 - 14 2208 77

View Full Product Details