Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pdc
Entrez Gene ID
20028
Gene Name
phosducin
Gene Aliases
Rpr-1, Rpr1
Chromosome Location
Chr.1: 150319417 - 150333906 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_024458.2 NP_077778.1 1 - 2 76 94
GenBank mRNA AK020798.1 - 1 - 2 77 94
BC012507.1 - 1 - 2 62 94
BC021920.1 - 1 - 2 77 94
L08075.1 - 1 - 2 95 94

View Full Product Details