Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a2
Entrez Gene ID
20526
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 2
Gene Aliases
AI266973, Glut-2, Glut2
Chromosome Location
Chr.3: 28697903 - 28728361 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031197.2 NP_112474.2 10 - 11 1481 68
GenBank mRNA AK005068.1 - 10 - 11 1482 68
AK149532.1 - 3 - 4 594 68
AK165430.1 - 10 - 11 1416 68
BC034675.1 - 10 - 11 1400 68
S77926.1 - 2 - 3 220 68
X15684.1 - 10 - 11 1430 68
X16986.1 - 10 - 11 1420 68

View Full Product Details