The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032002.2 | NP_114391.1 | 5 - 6 | 517 | 105 |
GenBank mRNA | AF083067.2 | - | 4 - 5 | 395 | 105 |
AK041077.1 | - | 6 - 7 | 549 | 105 | |
AK080058.1 | - | 5 - 6 | 469 | 105 | |
AK080161.1 | - | 7 - 8 | 611 | 105 | |
BC034839.1 | - | 8 - 9 | 810 | 105 |