The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053198.3 | NP_444428.3 | 5 - 6 | 356 | 75 |
GenBank mRNA | AF325263.1 | - | 5 - 6 | 298 | 75 |
BC089535.1 | - | 5 - 6 | 338 | 75 |