The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_054084.2 | NP_473425.2 | 1 - 2 | 80 | 79 |
GenBank mRNA | AK136518.1 | - | 1 - 2 | 74 | 79 |
BC111865.1 | - | 1 - 2 | 80 | 79 |