Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vps26a
Entrez Gene ID
30930
Gene Name
vacuolar protein sorting 26 homolog A (yeast)
Gene Aliases
AA407240, HB58, Vps26
Chromosome Location
Chr.10: 62454843 - 62486805 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113355.1 NP_001106826.1 7 - 8 942 102
NM_133672.3 NP_598433.1 7 - 8 869 102
GenBank mRNA AK028096.1 - 7 - 8 943 102
AK086851.1 - - 765 102
AK147989.1 - 7 - 8 810 102
AK149859.1 - 7 - 8 794 102
AK150244.1 - 7 - 8 767 102
AK151360.1 - 7 - 8 779 102
AK152424.1 - 7 - 8 813 102
AK159783.1 - 7 - 8 819 102
AK165853.1 - 7 - 8 813 102
AK167592.1 - 7 - 8 824 102
AK168213.1 - 7 - 8 869 102
AK168537.1 - 7 - 8 824 102
BC007148.1 - 7 - 8 790 102
BC058236.1 - 5 - 6 555 102
S41204.1 - 7 - 8 816 102

View Full Product Details