The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113355.1 | NP_001106826.1 | 8 - 9 | 1088 | 69 |
NM_133672.3 | NP_598433.1 | 8 - 9 | 1015 | 69 | |
GenBank mRNA | AK028096.1 | - | 8 - 9 | 1089 | 69 |
AK147989.1 | - | 8 - 9 | 956 | 69 | |
AK150244.1 | - | 8 - 9 | 913 | 69 | |
AK151360.1 | - | 8 - 9 | 925 | 69 | |
AK152424.1 | - | 8 - 9 | 959 | 69 | |
AK159783.1 | - | 8 - 9 | 965 | 69 | |
AK165853.1 | - | 8 - 9 | 959 | 69 | |
AK167592.1 | - | 8 - 9 | 970 | 69 | |
AK168213.1 | - | 8 - 9 | 1015 | 69 | |
AK168537.1 | - | 8 - 9 | 970 | 69 | |
BC007148.1 | - | 8 - 9 | 936 | 69 | |
BC058236.1 | - | 6 - 7 | 701 | 69 | |
S41204.1 | - | 8 - 9 | 962 | 69 |