The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133849.3 | NP_598610.1 | 2 - 3 | 671 | 83 |
NR_102309.1 | 2 - 3 | 667 | 83 | ||
GenBank mRNA | AK140374.1 | - | 2 - 3 | 676 | 83 |
AY044153.2 | - | 2 - 3 | 498 | 83 | |
AY142145.1 | - | 2 - 3 | 642 | 83 | |
BC132201.1 | - | 2 - 3 | 438 | 83 | |
BC145125.1 | - | 2 - 3 | 434 | 83 |