Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdk14
Entrez Gene ID
18647
Gene Name
cyclin-dependent kinase 14
Gene Aliases
Pftk1, mKIAA0834
Chromosome Location
Chr.5: 4803384 - 5380251 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011074.2 NP_035204.2 10 - 11 1165 83
GenBank mRNA AF033655.1 - 10 - 11 1124 83
AK083269.1 - 9 - 10 1320 83
AK087398.1 - 10 - 11 936 83
AK129226.1 - 10 - 11 992 83
AK138437.1 - 9 - 10 1406 83
AK139386.1 - 9 - 10 1326 83
AK141535.1 - 9 - 10 1094 83
AK165149.1 - 9 - 10 1010 83
BC068134.1 - 9 - 10 1369 83
U62391.1 - 9 - 10 1019 83

View Full Product Details