The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011137.3 | NP_035267.2 | 16 - 17 | 2082 | 70 |
NM_198932.3 | NP_945150.2 | 15 - 16 | 2010 | 70 | |
NM_198933.3 | NP_945151.2 | 15 - 16 | 2082 | 70 | |
NM_198934.3 | NP_945152.2 | 16 - 17 | 2154 | 70 | |
GenBank mRNA | AF508939.1 | - | 15 - 16 | 1991 | 70 |
AJ296212.1 | - | 15 - 16 | 2034 | 70 | |
AK028237.1 | - | 16 - 17 | 2084 | 70 | |
AK082573.1 | - | 3 - 4 | 745 | 70 | |
BC066201.1 | - | 15 - 16 | 1987 | 70 | |
S65461.1 | - | 14 - 15 | 1910 | 70 | |
X68362.1 | - | 16 - 17 | 2055 | 70 | |
X68363.1 | - | 15 - 16 | 1981 | 70 | |
X68364.1 | - | 15 - 16 | 1997 | 70 | |
X70325.1 | - | 5 - 6 | 823 | 70 |