The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146199.1 | NP_001139671.1 | 4 - 5 | 672 | 89 |
NM_011877.2 | NP_036007.2 | 4 - 5 | 807 | 89 | |
GenBank mRNA | BC014691.1 | - | 4 - 5 | 937 | 89 |
BC049893.1 | - | 3 - 4 | 639 | 89 | |
BC066856.1 | - | 3 - 4 | 610 | 89 | |
BC098500.1 | - | 4 - 5 | 672 | 89 | |
D37801.1 | - | 4 - 5 | 756 | 89 |