The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001199351.1 | NP_001186280.1 | 10 - 11 | 1088 | 79 |
NM_001199352.1 | NP_001186281.1 | 10 - 11 | 952 | 79 | |
NM_012040.3 | NP_036170.1 | 10 - 11 | 1028 | 79 | |
GenBank mRNA | AB023027.1 | - | 9 - 10 | 898 | 79 |
AF181984.1 | - | 10 - 11 | 1002 | 79 | |
AK158755.1 | - | 10 - 11 | 953 | 79 | |
BC051996.1 | - | 10 - 11 | 935 | 79 | |
BC055891.1 | - | 10 - 11 | 928 | 79 |